Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inositol, NF, 97-102%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Poly Bottle
Poly Drum
Poly Pail
87-89-8
C6H12O6
180.16
CDAISMWEOUEBRE-UHFFFAOYSA-N
cyclohexane-1,2,3,4,5,6-hexol
OC1C(O)C(O)C(O)C(O)C1O
87-89-8
C6H12O6
180.16
CDAISMWEOUEBRE-UHFFFAOYSA-N
cyclohexane-1,2,3,4,5,6-hexol
OC1C(O)C(O)C(O)C(O)C1O

I1012, 87-89-8, C6H12O6

Spectrum™ Inositol, NF
Packaging Poly Drum
Quantity 25 kg
CAS Max % 1
Percent Purity 97 to 102%