Learn More
Aviva Systems Biology Corp INPP5B Antibody - middle region (ARP54772_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54772_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against INPP5B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human INPP5B. Alternate names: 5PTase, MGC65156, MGC71303ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61307