Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INPP5F Mouse anti-Human, Mouse, Rat, Clone: S166A-26, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527672

Catalog No. PIMA527672


Add to cart

Description

Description

SAC2 is an inositol 1,4,5-trisphosphate (InsP3) 5-phosphatase and contains a Sac domain. The activity of this protein is specific for phosphatidylinositol 4,5-bisphosphate and phosphatidylinositol 3,4,5-trisphosphate. Alternatively spliced transcript variants have been observed, but most of them are not thought to be protein-coding.
Specifications

Specifications

INPP5F
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
D3ZGS3, Q01968, Q6NVF0
EC 3.1.3.36, Inositol polyphosphate 5 phosphatase OCRL 1, Inositol polyphosphate 5 phosphatase OCRL1, Inositol polyphosphate 5-phosphatase OCRL-1, INPP5F, LOCR, Lowe oculocerebrorenal syndrome protein, NPHL2, OCRL 1, OCRL, OCRL_HUMAN, OCRL1, Oculocerebrorenal syndrome of Lowe, Phosphatidylinositol polyphosphate 5 phosphatase
Mouse
IgG1
100 μg
-20°C
Primary
317576, 320634, 4952
Immunocytochemistry, Immunofluorescence, Western Blot
S166A-26
Unconjugated
OCRL
Liquid
Ocrl
Fusion protein amino acids 1-901 (full-length) of human INPP5F. Rat: 93% identity (845/904 amino acids identical). Mouse: 91% identity (824/901 amino acids identical) ∽50% identity with INPP5b.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit