Learn More
Echelon Biosciences Research Labs Ins(1,4,5)P3 BIOTIN 10ug
SDP
Supplier:  Echelon Biosciences Research Labs Q145B10UG
Encompass
Inositol phosphates are important signaling molecules involved in numerous cellular functions. This Inositol 1,4,5-trisphosphate (Ins(1,4,5)P3) analog is labeled with biotin through a three-carbon linker at the 1-position on the inositol ring. This item is non returnable
Catalog No. 50-143-996