Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INS Mouse anti-Human, Clone: INS04 + INS05, Abnova™

Mouse monoclonal antibody raised against human INS.

Manufacturer:  Abnova Corporation MAB12631

Catalog No. 89-934-378


Add to cart

Description

Description

After removal of the precursor signal peptide, proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds. Binding of this mature form of insulin to the insulin receptor (INSR) stimulates glucose uptake. A variety of mutant alleles with changes in the coding region have been identified. There is a read-through gene, INS-IGF2, which overlaps with this gene at the 5' region and with the IGF2 gene at the 3' region. [provided by RefSeq]

Specifications

Specifications

INS
INS04 + INS05
Mouse monoclonal antibody raised against human INS.
In 10mM PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
ILPR/IRDN
Mouse
Protein G purification
RUO
Primary
3630
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
INS
Human INS
50 ug
Store at 4°C. Do not freeze.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit