Learn More
Thermo Scientific™ InSight Wireless Monitoring Solution Kits
Designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events
Manufacturer:  Thermo Scientific™ KITT3C1D2WIF1002
 View more versions of this product
Catalog No. 11-676-510
Add to cart
Description
Description
Monitors:
  • Cabinet temperature
  • Electrical use and consumption
  • Door openings
  • Ambient temperature
Features:
  • Powered by KLATU™
  • Monitors 24/7
  • Alerts when threshold exceeded
  • Data run through InSight software
  • Compare asset performance against industry standards
Data collection
  • Sent wirelessly from the InSight mote via your Wi-Fi network to InSight gateway server
  • Collects and transmits to Cloud storage
  • Wireless 915MHZ systems (North America only) are also available for smaller installations (25 motes or less)
  • Analysis provides real-time comparisons for knowledge-based decisions
  • Warranty varies by country, please contact us for details
Specifications
Specifications
No CFR part 11 compliance
ABS plastic
3.3V
18–24 hours. Full recovery from network failure.
LED (2) with various sensor node status messages
Accuracy: RTD Sensor- ±0.3°C; Thermocouple Sensor - ±1.5°C (at -100ºC)
50 amperes maximum
5.4 x 2.5 x 1.1 in. (13.8 x 6.3 x 2.7 cm)
Includes PT100, current transducer and door switch. Also includes 1 asset license; InSight refigerator license
RTD Sensor: –200°C to +600°C; Thermocouple Sensor: -200°C to +200°C
802.11(n) for refrigerators
Cabinet temperature, electrical use and energy consumption, door openings and ambient temperature