Learn More
Abnova™ INSR (Human) Recombinant Protein
For use with Functional Study.
Supplier: Abnova™ P6505
Description
Human INSR (NP_000199.1, 1005 a.a. - 1310 a.a.) partial recombinant protein with GST-tag at N-terminal using baculovirus expression system.Specifications
Functional Study | |
insulin receptor | |
5 μg | |
CD220, HHF5 |
3643 | |
The purity was assessed by SDS-PAGE/CBB staining. | |
Stored at -80°C. Aliquot to avoid repeated freezing and thawing. |