Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Instant Buffer S70 pH7.0 (acc. to Soerensen) (50packs), TCI America™
Click to view available options
Quantity:
SET
Specifications
Color | White |
Quantity | SET |
MDL Number | MFCD00147253 |
Physical Form | Granul at 20°C |
Chemical Name or Material | Instant Buffer S70 pH7.0 (acc. to Soerensen) (50packs) |
Safety and Handling
TSCA : No