Learn More
Sklar InstrumentsSupplier Diversity Partner Instrument Cleaning Brush, Sklar, Nylon / Stainless Steel. This product is 25mm x 15 inches. Pack of 24. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101672
Instrument Cleaning Brush, Sklar, Nylon / Stainless Steel. Cannula Instrument Cleaning Brushes are used in the multiple-step process of sterilizing a reusable cannula. Following soaking in enzymatic solution, the cannula should be cleaned manually with a brush and detergent or ultrasonically. This product is 25mm x 15 inches. Pack of 24.
Catalog No. 50-143-7085