Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Instrument-Specific Serum Filters

Catalog No. 0268155 Shop All Fisherbrand Products
Change view
Click to view available options
Length (English):
1.25 in.
1.37 in.
1.87 in.
2.25 in.
2.5 in.
Length (Metric):
31.75mm
34.7mm
47mm
57mm
63mm
Diameter (English) Outside:
0.5 in.
0.6 in.
Diameter (Metric) Outside:
13mm
16mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Length (Metric) Diameter (English) Outside Diameter (Metric) Outside
02-681-55 2.5 in. 63mm 0.6 in. 16mm
02-681-56 2.25 in. 57mm 0.5 in. 13mm
02-681-61 1.87 in. 47mm 0.6 in. 16mm
02-681-60 1.37 in. 34.7mm 0.5 in. 13mm
02-681-59 1.25 in. 31.75mm 0.6 in. 16mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-681-55
Only null left
Add to Cart
Add to Cart

Designed to remove fibrin and gel separation particulates and permit direct sampling from primary collection tubes.

  • Help protect ion selective electrodes (ISE) and eliminate additional caps, tubes and specimen labeling
  • Use with glass or plastic collection tubes

  • Technique reduces second centrifugation, tube labeling and sample preparation time
  • Allow direct sampling into automatic laboratory equipment
  • Eliminate fibrin, particulate matter and equipment downtime

Specifications

Diameter (English) Outside 0.6 in.
For Use With (Equipment) Collection tube 16 x 100mm
Length (Metric) 63mm
Diameter (Metric) Outside 16mm
Quantity 100 Pack
Length (English) 2.5 in.
Product Type Serum Filter
For Use With (Application) Remove fibrin and gel separation particulates and permit direct sampling from primary collection tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.