Learn More
Description
- Help protect ion selective electrodes (ISE) and eliminate additional caps, tubes and specimen labeling
- Use with glass or plastic collection tubes
- Technique reduces second centrifugation, tube labeling and sample preparation time
- Allow direct sampling into automatic laboratory equipment
- Eliminate fibrin, particulate matter and equipment downtime
Specifications
Specifications
| Diameter (English) Outside | 0.6 in. |
| Length (English) | 1.25 in. |
| Quantity | 100 Pack |
| Diameter (Metric) Outside | 16mm |
| For Use With (Equipment) | Collection tube 16 x 75mm |
| Length (Metric) | 31.75mm |
| Product Type | Serum Filter |
| For Use With (Application) | Remove fibrin and gel separation particulates and permit direct sampling from primary collection tubes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.