Learn More
Fisherbrand™ Instrument-Specific Serum Filters
Designed to remove fibrin and gel separation particulates and permit direct sampling from primary collection tubes.
$101.00 - $111.00
Specifications
Product Type Serum Filter
For Use With (Application) Remove fibrin and gel separation particulates and permit direct sampling from primary collection tubes
Quantity 100 Pack
View More Specs
Products 5
Catalog Number Diameter (English) Outside Diameter (Metric) Outside Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Diameter (English) Outside Diameter (Metric) Outside Length (English) Length (Metric) Price Quantity & Availability  
02-681-56
Accessories View Documents
0.5 in. 13mm 2.25 in. 57mm
Pack of 100 for $111.00
Case of 40 PK for $3,571.50
Add to cart
02-681-61
Accessories View Documents
0.6 in. 16mm 1.87 in. 47mm
Pack of 100 for $111.00
Case of 40 PK for $3,579.98
Add to cart
02-681-55
Accessories View Documents
0.6 in. 16mm 2.5 in. 63mm
Pack of 100 for $111.00
Case of 40 PK for $3,580.50
Add to cart
02-681-60
Accessories View Documents
0.5 in. 13mm 1.37 in. 34.7mm
Pack of 100 for $111.00
Case of 40 PK for $3,556.30
Add to cart
02-681-59
Accessories View Documents
0.6 in. 16mm 1.25 in. 31.75mm
Pack of 100 for $101.00
Case of 40 PK for $3,247.00
Add to cart
Description
Description
  • Help protect ion selective electrodes (ISE) and eliminate additional caps, tubes and specimen labeling
  • Use with glass or plastic collection tubes
  • Technique reduces second centrifugation, tube labeling and sample preparation time
  • Allow direct sampling into automatic laboratory equipment
  • Eliminate fibrin, particulate matter and equipment downtime
Specifications
Specifications
Serum Filter
Remove fibrin and gel separation particulates and permit direct sampling from primary collection tubes
100 Pack