Learn More
Sklar InstrumentsSupplier Diversity Partner Instrument Tray, Sklar, Solid, Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. The overall size of this tray is 16-1/2 inches x 10 inches x 2-1/2 inches. Pack of 12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 10174812
Instrument Tray, Sklar, Solid, Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this tray is 16-1/2 inches x 10 inches x 2-1/2 inches. Pack of 12.
Catalog No. 50-143-7118