Learn More
Sklar InstrumentsSupplier Diversity Partner Instrument Tray Cover, Sklar, 6-7/8 in., Sklar's Instrument Tray Covers are are seamless to prevent bacterial growth. This flat lid is compatible with tray 10-1755. It measures 6-7/8 inches x 4-1/4 inches and is sold in packs of 6. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 1017376
Instrument Tray Cover, Sklar, 6-7/8 in., Sklar's Instrument Tray Covers are are seamless to prevent bacterial growth. This flat lid is compatible with tray 10-1755. It measures 6-7/8 inches x 4-1/4 inches and is sold in packs of 6.
Catalog No. 50-143-7111