Learn More
Sklar InstrumentsSupplier Diversity Partner Instrument Tray, Sklar, 20-3/4 in. x 12-3/4 in. x 6 in., Stainless Steel. Sklar Instrument . PK/6. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 1019486
Instrument Tray, Sklar, 20-3/4 in. x 12-3/4 in. x 6 in., Stainless Steel. Sklar Instrument Trays are used for sterilizing, storing and transporting instruments for surgery. Available in a variety of sizes, they feature a seamless design to prevent bacterial growth. Dome or flat lids are also available. The overall size of this tray is 20-3/4 inches x 12-3/4 inches x 6 inches. Pack of 6.
Catalog No. 50-143-7155