Learn More
Vernier™ Instrumentation Amplifier Products for Science Education
The Instrumentation Amplifier can be used to connect a wide variety of measurement equipment to a Vernier interface and electronically collect, store, and analyze the data.
Manufacturer:  Vernier™ INABTA
Catalog No. S16176ND
Add to cart
Includes: Vernier Instrumentation Amplifier
Description
Description
Gain settings: 150, 75, 7.5, 15, 3, and 1.5±5% Linearity: 1% Power: 2.5 mA @ 5 VDC Frequency Response: 0-10 kHz (f3dB) Impedance: 1 MΩ to ground Range settings: 0-20 mV, 0-200 mV, 0-1 V, ±20 mV, ±200 mV, ±1 V
Warranty and Services
5-year limited warranty
Specifications
Specifications
5-year limited warranty
Educational use only
Required
Education
High School and College
0 to 10 kHz (f3dB) Frequency Response