Learn More
Cole-Parmer Megger MIT220-EN 250V/500V Insulation and Continuity Tester  DFS Item
Manufacturer:  Cole-Parmer 2002248
The MIT200 series insulation and continuity testers are ideal for testing transformers, motors, generators, switchgear, panel building, domestic appliances, power tools, etc., as well as fixed electrical wiring systems. Their small size and light weight make them ideal for those engineers that need to carry them for extended periods.
Catalog No. 50-631-598