Learn More
MP Biomedical™ Insulin ChLIA Kit
Quantitative Chemiluminescence Immunoassay
Manufacturer:  MP Biomedical™ 07M2475A
Catalog No. MP07M2475A
Add to cart
Description
Description
The Insulin Chemiluminescence Immunoassay kit is intended for the measurement of Insulin concentration in human serum.
Specifications
Specifications
Kit and Assay
50 μL
1 x 96 Wells
Serum
FDA Exempt
2°C to 8°C
Human