Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Mouse anti-Human, Mouse, Alexa Fluor™ 350, Clone: 2D11-H5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP233146AF350

Catalog No. NP233146A30


Add to cart

Description

Description

Insulin Monoclonal antibody specifically detects Insulin in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

Insulin
2D11-H5
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Conjugated, Purified
INS
The antibody reacts with the Insulin antigen.
Protein A or G purified
Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers
Primary
3630.0
Human, Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Insulin is a protein consisting of an alpha-chain of 21 amino acids and a beta-chain of 30 amino acids and produced in the beta-cells of the pancreas. 2D11-H5 is a specific insulin mAb as tested by ELISA and on human pancreatic tissue. Source: A BALB/c mouse was immunised with swine insulin. The mouse spleen cells were fused with SP2/0 Ag14 cells and the clones were selected on anti-insulin activity.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit