Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulin R/CD220, Rabbit anti-Human, Clone: 83-7, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP2810390.025MG
Description
Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Insulin R/CD220 | |
Monoclonal | |
1 mg/mL | |
ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin | |
CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Recognises the extracellular domain of the Insulin R/CD220. Amino acids 469 - 592 (exon 7 and 8) of Insulin R/CD220. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG κ |
ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
83-7 | |
Unconjugated | |
PBS | |
INSR | |
Epitope within amino acids 140-301 (the cysteine rich region) of the extracellular domain of the human Insulin R/CD220. | |
0.025 mg | |
Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
3643 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction