Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Rabbit anti-Human, Mouse, Porcine, Bovine, Rabbit, Clone: IRDN/1980R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Insulin
Clone IRDN/1980R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122496
View Documents
Novus Biologicals
NBP30751120UG
20 μg
Each for $159.82
Add to cart
 
NB122495
View Documents
Novus Biologicals
NBP30751110UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

Insulin Monoclonal antibody specifically detects Insulin in Human, Mouse, Porcine, Bovine, Rabbit samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Insulin
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human, Mouse, Porcine, Bovine, Rabbit
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Recombinant human full-length Insulin protein (IRDN)
Monoclonal
Protein A or G purified
IRDN/1980R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers
10 mM PBS with 0.05% BSA
3630
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit