Learn More
Insulin Receptor (α-subunit) Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ Insulin Receptor (α-Subunit) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
Supplier: Epredia MS632P1ABX
Description
The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain, whereas the α-subunit (135kDa) is completely extracellular.Host Species: Mouse
Clone: 83-7
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Pig, Sheep and Rabbit. Does not react with mouse and rat.
Epitope: Exon 3
Immunogen: IM-9 lymphocytes followed by IR
Molecular Weight: 135kDa
Positive Control: IM-9 lymphocyte cells, Placenta
Cellular Localization: Cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- ELISA
- Flow Cytometry
- Insulin-like Agonist
- Does Not Significantly Inhibit Insulin Binding
- Tyrosine Kinase Assay (Ab-mediated capture on microplates)
Note:
USA: RUO; Int'l: RUO
Specifications
Insulin Receptor (alpha-Subunit) Ab-2 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Steroids Hormones and Growth Factors | |
Bovine, Human, Porcine, Rabbit, Sheep | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
83-7 | |
Unconjugated | |
IM-9 lymphocytes followed by IR | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.