Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Insulin Receptor (α -Subunit) Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in experiments with Thermo Scientific™ Insulin Receptor (α-Subunit) Ab-2, Mouse Monoclonal Antibody.

$161.00 - $970.00

Specifications

Antigen Insulin Receptor (alpha-Subunit) Ab-2
Clone 83-7
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Murine
View More Specs

Products 10
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS632B
View Documents
Thermo Scientific™
MS632B
Biotinylated Ab with BSA and Azide 1mL
Each for $970.00
Add to cart
 
MS632B0
View Documents
Thermo Scientific™
MS632B0
Biotinylated Ab with BSA and Azide 100μL
Each for $225.50
Add to cart
 
MS632B1
View Documents
Thermo Scientific™
MS632B1
Biotinylated Ab with BSA and Azide 500μL
Each for $615.50
Add to cart
 
MS632P
View Documents
Thermo Scientific™
MS632P
Purified Ab with BSA and Azide 1mL
Each for $785.50
Add to cart
 
MS632P0
View Documents
Thermo Scientific™
MS632P0
Purified Ab with BSA and Azide 100μL
Each for $161.00
Add to cart
 
MS632P1
View Documents
Thermo Scientific™
MS632P1
Purified Ab with BSA and Azide 500μL
Each for $599.00
Add to cart
 
MS632P1ABX
View Documents
Thermo Scientific™
MS632P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $550.50
Add to cart
 
MS632PABX
View Documents
Thermo Scientific™
MS632PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $837.00
Add to cart
 
MS632R7
View Documents
Thermo Scientific™
MS632R7
Ready-to-use for IHC (UVLP) 7mL
Each for $665.50
Add to cart
 
MS632R1
View Documents
Thermo Scientific™
MS632R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in experiments with Thermo Scientific™ Insulin Receptor (α-Subunit) Ab-2, Mouse Monoclonal Antibody.

The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain, whereas the α-subunit (135kDa) is completely extracellular.

Host Species: Mouse

Clone: 83-7

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Pig, Sheep and Rabbit. Does not react with mouse and rat.

Epitope: Exon 3

Immunogen: IM-9 lymphocytes followed by IR

Molecular Weight: 135kDa

Positive Control: IM-9 lymphocyte cells, Placenta

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • ELISA
  • Flow Cytometry
  • Insulin-like Agonist
  • Does Not Significantly Inhibit Insulin Binding
  • Tyrosine Kinase Assay (Ab-mediated capture on microplates)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Insulin Receptor (alpha-Subunit) Ab-2
Immunohistochemistry (Paraffin)
Murine
Steroids Hormones and Growth Factors
IM-9 lymphocytes followed by IR
83-7
Monoclonal
RUO
Primary
IgG2a kappa
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit