Learn More
Insulin Receptor (β-subunit) Ab-6 Mouse Monoclonal Antibody, Epredia™
Epredia™ Insulin Receptor (β-Subunit) Ab-6, Mouse Monoclonal Antibody provides accurate, reproducible results.
$304.00 - $1086.00
Specifications
Antigen | Insulin Receptor (beta-Subunit) Ab-6 |
---|---|
Clone | CT-3 |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS636P0
|
Epredia
MS636P0 |
Concentrated | 100 μL |
Each for $304.00
|
|
||||
MS636P1
|
Epredia
MS636P1 |
Concentrated | 500 μL |
Each for $828.00
|
|
||||
MS636P
|
Epredia
MS636P |
Concentrated | 1 mL |
Each for $1,086.00
|
|
||||
MS636P1ABX
|
Epredia
MS636P1ABX |
Concentrated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS636PABX
|
Epredia
MS636PABX |
Concentrated | 200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS636R1
|
Epredia
MS636R1 |
Prediluted | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS636R7
|
Epredia
MS636R7 |
Prediluted | 7 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain with tyrosine kinase acivity, whereas the α-subunit (135kDa) is completely extracellular.Host Species: Mouse
Clone: CT-3
Isotype: IgG1
Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.
Epitope: C-terminal
Immunogen: Recombinant fragment including the C-terminal 100 amino acids of human insulin receptor
Molecular Weight: 95kDa
Positive Control: HEP-G-2, IM-9 lymphocyte cells; Placenta or brain
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Not suitable)
- Affinity Purification (With peptide elution)
- ELISA (As capture antibody only)
- Tyrosine Kinase Assay (Ab-mediated capture on microplates)
Note:
USA: RUO; Int'l: RUO
Specifications
Insulin Receptor (beta-Subunit) Ab-6 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Non-human Primate, Rat | |
IgG1 |
CT-3 | |
Monoclonal | |
Mouse | |
Steroids Hormones and Growth Factors | |
Recombinant-fragment including the C-terminal 100 amino acids of human insulin receptor | |
Primary |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.