Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Insulin R/CD220 Rabbit anti-Human, Mouse, Rat, Clone: SN20-13, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267726
Description
Insulin R/CD220 Monoclonal antibody specifically detects Insulin R/CD220 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
Insulin R/CD220 | |
SN20-13 | |
Unconjugated | |
Purified | |
CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
INSR | |
Recombinant Protein | |
Monoclonal | |
Protein A purified | |
Diabetes Research, Lipid and Metabolism, Neuroscience, Nutrient Sensing in the Brain, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
Human,Mouse,Rat |
Immunocytochemistry,Immunofluorescence,Western Blot | |
1 mg/mL | |
Western Blot 1:1000-1:2000 Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
3643 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title