Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INTS10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550338

Catalog No. PA550338


Add to cart

Description

Description

The antibody detects endogenous levels of total INT10 protein.

INTS10 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates 3-prime end processing of small nuclear RNAs U1.
Specifications

Specifications

INTS10
Polyclonal
Unconjugated
INTS10
Liquid
INTS10
Fusion protein corresponding to residues near the C terminal of human integrator complex subunit 10
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1.9 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9NVR2
C8orf35
Rabbit
IgG
100 μL
-20°C
Primary
55174
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit