Learn More
Bevco™ Integra ESD/Cleanroom Vinyl Chair
Bevco's Integra ESD/Cleanroom ISO Certified Upholstered Seating combines long-lasting comfort and durability with a heavy-duty polypropylene back shell to protect the upholstery. A generous seat size and high-density foam provide an exceptional seating experience and long-lasting comfort.
$457.00 - $627.00
Specifications
Adjustable Yes
Adjustable (Height) Yes
Quantity 1
Back Style Ergonomic
Base Style Polished Aluminum
View More Specs
Products 14
Catalog Number Mfr. No. Certifications Seating Type Color Price Quantity & Availability  
Catalog Number Mfr. No. Certifications Seating Type Color Price Quantity & Availability  
19-058-256
View Documents
Bevco™
9050ME1BLK
ISO4 Certified Desk Height Black
Each for $542.00
Add to cart
19-058-258
View Documents
Bevco™
9550ME1BLK
ISO4 Certified High Bench Height Black
Each for $627.00
Add to cart
19-058-260
View Documents
Bevco™
9050ME2BLK
ISO5 Certified Desk Height Black
Each for $505.00
Add to cart
19-058-262
View Documents
Bevco™
9550ME2BLK
ISO5 Certified High Bench Height Black
Each for $577.00
Add to cart
19-058-264
View Documents
Bevco™
9050ME2BLU
ISO5 Certified Desk Height Blue
Each for $505.00
Add to cart
19-058-266
View Documents
Bevco™
9550ME2BLU
ISO5 Certified High Bench Height Blue
Each for $577.00
Add to cart
19-058-268
View Documents
Bevco™
9050ME3BLK
ISO6 Certified Desk Height Black
Each for $481.00
Add to cart
19-058-271
View Documents
Bevco™
9550ME3BLK
ISO6 Certified High Bench Height Black
Each for $553.00
Add to cart
19-058-273
View Documents
Bevco™
9050ME3BLU
ISO6 Certified Desk Height Blue
Each for $481.00
Add to cart
19-058-275
View Documents
Bevco™
9550ME3BLU
ISO6 Certified High Bench Height Blue
Each for $553.00
Add to cart
19-058-278
View Documents
Bevco™
9050ME4BLK
ISO7 Certified Desk Height Black
Each for $457.00
Add to cart
19-058-281
View Documents
Bevco™
9550ME4BLK
ISO7 Certified High Bench Height Black
Each for $529.00
Add to cart
19-058-283
View Documents
Bevco™
9050ME4BLU
ISO7 Certified Desk Height Blue
Each for $457.00
Add to cart
19-058-285
View Documents
Bevco™
9550ME4BLU
ISO7 Certified High Bench Height Blue
Each for $529.00
Add to cart
Description
Description
  • Sturdy construction and modern design makes this is a great seating solution for critical rooms
  • Static dissipative when measured to the ANSI/ESD STM12.1-2019 Standards
  • Designed to meet the particulate goals of ISO 14644-1
  • Options of ISO 4, 5, 6, and 7 are available to meet all cleanroom needs
  • A generous seat size provides an exceptional seating experience
  • Thick, resilient, high-density foam provides long-lasting comfort
  • Specifically shaped backrest provides full back support and comes standard with a heavy-duty polypropylene back shell to protect the upholstery
  • Easy-to-use pneumatic seat height adjustment with lifetime cylinder guarantee
  • Sturdy, 5-legged base ensures stability at any height range
  • ESD vinyl meets anti-flammability requirements of California Technical Bulletin 117
  • Dual-wheel ESD hard floors casters are available (adds one inch to seat height)
  • Meets ANSI/BIFMA standards
  • GreenGuard Gold certified
  • Supports up to 300 lb.
  • 12 year warranty
Specifications
Specifications
Yes
1
Polished Aluminum
18 in.
3 in.
Vinyl
Yes
Yes
Yes
Ergonomic
300 lb.
Vinyl
3 in.
No
No