Integra™ Miltex™ EZ-Zyme™ All-Purpose Enzyme Cleaner
All-purpose neutral pH all purpose multi-enzyme concentrate
Manufacturer: Integra™ Miltex™ 3755
 View more versions of this product
Catalog No. 12-460-427
  Check Availability
Add to Cart
    Specifications
    Size 1 gal.
    Container Type Pump bottle
    For Use With (Application) All purpose neutral pH all purpose multi enzyme concentrate, holding and presoak, soak cleaning, ultrasonic cleaning, manual cleaning, laundry presoak
  • Description & Specifications
Related Products
Product Content Feedback