Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Goldstein Retractor

3 x 3 pointed prongs

Manufacturer:  Integra™ Miltex™ 1888

Catalog No. 12-460-330

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • High-quality surgical-grade stainless steel
  • 5mm deep
  • Length: 1.125 in. (2.8cm)

Specifications

Specifications

Premium
2.8 cm
Goldstein Retractor
1.12 in.
Stainless Steel
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit