Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integra™ Miltex™ Magnifying Loupes
Quickly and easily locked into personal setting using a unique locking knob
Supplier: Integra™ Miltex™ 15005
This item is not returnable.
View return policy
Description
- Dual hinge design
- Locking interpupillary knob
- Double locking convergence angle
Specifications
Loupe, Magnifying | |
6.35 cm | |
Red | |
For endodontic procedures | |
Regular |
33 to 40.64 cm | |
2.5 in. | |
3x | |
13 to 16 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction