Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Mayo Scissors

Offer outstanding distal cutting capability

$308.00 - $829.00

Specifications

Grade Premium
Material Stainless Steel
For Use With (Application) Dissecting
View More Specs

Products 14
Catalog Number Mfr. No. Blade Style Length (English) Length (Metric) Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Blade Style Length (English) Length (Metric) Product Type Price Quantity & Availability  
12-460-467
View Documents
Integra™ Miltex™
5122
Curved, Beveled 5.5 in. 13.97 cm Mayo Scissors
Each for $327.00
Add to Cart
 
12-460-466
View Documents
Integra™ Miltex™
5120
Straight, Beveled 5.5 in. 13.97 cm Mayo Scissors
Each for $308.00
Add to Cart
 
12-460-468
View Documents
Integra™ Miltex™
5122TC
Curved, Beveled 5.5 in. 13.97 cm Carb-N-Sert Mayo Scissors
Each for $712.00
Add to Cart
 
12-460-471
View Documents
Integra™ Miltex™
5126
Curved, Beveled 6.75 in. 17.14 cm Mayo Scissors
Each for $408.00
Add to Cart
 
12-460-469
View Documents
Integra™ Miltex™
5124
Straight, Beveled 6.75 in. 17.14 cm Mayo Scissors
Each for $379.00
Add to Cart
 
12-460-470
View Documents
Integra™ Miltex™
5124TC
Straight, Beveled 6.75 in. 17.14 cm Carb-N-Sert Mayo Scissors
Each for $640.00
Add to Cart
 
12-460-472
View Documents
Integra™ Miltex™
5128
Straight, Beveled 9 in. 22.86 cm Mayo Scissors
Each for $547.00
Add to Cart
 
12-460-473
View Documents
Integra™ Miltex™
5136
Straight, Rounded Tips 5.5 in. 13.97 cm Mayo Scissors
Each for $435.00
Add to Cart
 
12-460-474
View Documents
Integra™ Miltex™
5136TC
Straight, Rounded Tips 5.5 in. 13.97 cm Carb-N-Sert Mayo Scissors
Each for $642.00
Add to Cart
 
12-460-475
View Documents
Integra™ Miltex™
5140TC
Straight, Rounded Tips 6.75 in. 17.14 cm Carb-N-Sert Mayo Scissors
Each for $339.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
12-460-506
View Documents
Integra™ Miltex™
5SC120
Straight, Beveled 5.5 in. 13.97 cm SuperCut Mayo Scissors
Each for $611.00
Add to Cart
 
12-460-508
View Documents
Integra™ Miltex™
5SC124
Straight, Beveled 6.75 in. 17.14 cm SuperCut Mayo Scissors
Each for $592.00
Add to Cart
 
12-460-507
View Documents
Integra™ Miltex™
5SC122
Curved, Beveled 5.5 in. 13.97 cm SuperCut Mayo Scissors
Each for $563.00
Add to Cart
 
12-460-509
View Documents
Integra™ Miltex™
5SC128
Straight, Beveled 9 in. 22.86 cm SuperCut Mayo Scissors
Each for $829.00
Add to Cart
 
Description

Description

  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
  • Sharp/sharp scissors available in straight and curved patterns
  • Carb-N-Sert™ scissors have tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
  • SuperCut scissors have a specially designed razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to prevent tissue slippage

Order Info

Cold or chemical sterilization not recommended for Carb-N-Sert scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-shapr edge to nick or become irregular.

Specifications

Specifications

Premium
Stainless Steel
Dissecting
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit