Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Vannas Capsulotomy Scissors

For intraocular, ear, nose and throat procedures

$850.00 - $873.00

Specifications

For Use With (Application) Surgical, ophthalmic
Grade Premium
Material Stainless Steel
Type Vannas Capsulotomy Scissors
View More Specs

Products 2
Catalog Number Mfr. No. Blade Style Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Blade Style Length (English) Length (Metric) Price Quantity & Availability  
12-460-283
View Documents
Integra™ Miltex™
181623
Straight 3.25 in. 8.3cm
Each for $850.00
Add to cart
 
12-460-282
View Documents
Integra™ Miltex™
181622
Curved 3.125 in. 7.9cm
Each for $873.00
Add to cart
 
Description

Description

  • Surgical-grade stainless steel
  • Fully tempered screw locks maintains alignment
  • Design combines weight, balance and feel for quality performance
  • Well rounded and polished ring handles
  • Extra delicate
  • Sharp points

Specifications

Specifications

Surgical, ophthalmic
Premium
Stainless Steel
Vannas Capsulotomy Scissors
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit