Learn More
Bevco™ Integra Tiltable ESD/Cleanroom Vinyl Chair
Bevco's Integra ESD/Cleanroom ISO Certified Upholstered Seating combines long-lasting comfort and durability with a heavy-duty polypropylene back shell to protect the upholstery. A generous seat size and high-density foam provide an exceptional seating experience and long-lasting comfort.
$505.00 - $730.00
Specifications
Adjustable Yes
Adjustable (Height) Yes
Quantity 1
Back Style Ergonomic
Base Style Polished Aluminum
View More Specs
Products 14
Catalog Number Mfr. No. Certifications Color Seating Type Price Quantity & Availability  
Catalog Number Mfr. No. Certifications Color Seating Type Price Quantity & Availability  
19-058-257
View Documents
Bevco™
9051ME1BLK
ISO4 Certified Black Desk Height
Each for $631.00
Add to cart
19-058-259
View Documents
Bevco™
9551ME1BLK
ISO4 Certified Black High Bench Height
Each for $730.00
Add to cart
19-058-261
View Documents
Bevco™
9051ME2BLK
ISO5 Certified Black Desk Height
Each for $569.00
Add to cart
19-058-263
View Documents
Bevco™
9551ME2BLK
ISO5 Certified Black High Bench Height
Each for $654.00
Add to cart
19-058-265
View Documents
Bevco™
9051ME2BLU
ISO5 Certified Blue Desk Height
Each for $569.00
Add to cart
19-058-267
View Documents
Bevco™
9551ME2BLU
ISO5 Certified Blue High Bench Height
Each for $654.00
Add to cart
19-058-269
View Documents
Bevco™
9051ME3BLK
ISO6 Certified Black Desk Height
Each for $529.00
Add to cart
19-058-272
View Documents
Bevco™
9551ME3BLK
ISO6 Certified Black High Bench Height
Each for $601.00
Add to cart
19-058-274
View Documents
Bevco™
9051ME3BLU
ISO6 Certified Blue Desk Height
Each for $529.00
Add to cart
19-058-277
View Documents
Bevco™
9551ME3BLU
ISO6 Certified Blue High Bench Height
Each for $601.00
Add to cart
19-058-279
View Documents
Bevco™
9051ME4BLK
ISO7 Certified Black Desk Height
Each for $505.00
Add to cart
19-058-282
View Documents
Bevco™
9551ME4BLK
ISO7 Certified Black High Bench Height
Each for $577.00
Add to cart
19-058-284
View Documents
Bevco™
9051ME4BLU
ISO7 Certified Blue Desk Height
Each for $505.00
Add to cart
19-058-286
View Documents
Bevco™
9551ME4BLU
ISO7 Certified Blue High Bench Height
Each for $577.00
Add to cart
Description
Description
  • Sturdy construction and modern design makes this is a great seating solution for critical rooms
  • Static dissipative when measured to the ANSI/ESD STM12.1-2019 Standards
  • Designed to meet the particulate goals of ISO 14644-1
  • Options of ISO 4, 5, 6, and 7 are available to meet all cleanroom needs
  • A generous seat size provides an exceptional seating experience
  • Thick, resilient, high-density foam provides long-lasting comfort
  • Specifically shaped backrest provides full back support and comes standard with a heavy-duty polypropylene back shell to protect the upholstery
  • Easy-to-use pneumatic seat height adjustment with lifetime cylinder guarantee
  • Sturdy, 5-legged base ensures stability at any height range
  • ESD vinyl meets anti-flammability requirements of California Technical Bulletin 117
  • Dual-wheel ESD hard floors casters are available (adds one inch to seat height)
  • Meets ANSI/BIFMA standards
  • GreenGuard Gold certified
  • Supports up to 300 lb.
  • 12 year warranty
Specifications
Specifications
Yes
1
Polished Aluminum
18 in.
3 in.
Vinyl
Yes
Yes
Yes
Ergonomic
300 lb.
Vinyl
3 in.
No
No