Learn More
3M™ Integrated Protective Eyewear
3M™ Integrated Protective Eyewear is lightweight retractable eyewear that is easy to attach to the headband of 3M hard hats
Manufacturer:  3M™ 7010342442
 View more versions of this product
Catalog No. 19-033-501
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Attaches directly to hard hat suspension, nearly eliminating the weight of the eyewear on the nose and temples
  • Retracts between hard hat suspension and shell, helping to protect the lens when not in use
  • Stable, stays in place and secure when not in use
  • Easy to maneuver and adjust for closeness and height
  • Comfort: No temples, minimal weight on nose bridge
  • Anti-fog lens helps prevent moisture from building up on the lens
  • Connects directly to the ratchet and pinlock suspension systems of the 3M™ H-700 and 3M™ H-800 Series of hard hats
Specifications
Specifications
Anti-fog
Amber
ANSI Z89.1 2010
Integrated Protective Eyewear
3M™ H 700 and H 800 Series of hard hats, 3M™ SecureFit™ Safety Helmet X5000
Polycarbonate
HIE602AF
20/Cs.
Assembly, Grinding, Machine Operations, Painting, Remediation, Sanding, Welding
Anti-fog