Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin β1 Rabbit anti-Human,Monkey,Mouse,Rat, KHL, Clone: 1A27, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Integrin β1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB123025MI
View Documents
MilliporeSigma
ZRB123025UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB12304XMI
View Documents
MilliporeSigma
ZRB12304X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Integrin β1 Monoclonal antibody specifically detects Integrin β1 in Human,Monkey,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

Integrin β1
KHL
Rabbit
Fibronectin receptor subunit beta;Glycoprotein IIa;GPIIA;VLA-4 subunit beta;CD29
KLH-conjugated linear peptide corresponding to 23 amino acids from the C-terminal, cytoplasmic domain of human Integrin beta1, isoform 1.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Monkey, Mouse, Rat
ITGB1, FNRB, MDF2, MSK12
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit