Learn More
Integrin alpha 10 Rat anti-Mouse, Alexa Fluor 405, Clone: 885501, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB9757V100UG
Catalog No. FAB9757V100
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Integrin alpha 10
885501
Flow Cytometry 0.25-1 ug/10^6 cells
NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1)
integrin alpha-10, integrin, alpha 10, ITGA10, PRO827
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 10 beta 1 Phe23-Thr1119 (Integrin alpha 10) and Gln21-Asp728 (Integrin beta 1) Accession # NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1)
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse Integrin alpha 10 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Intagrin alpha 2 beta 1 was observered.
Flow Cytometry
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Supernatant
Rat
IgG2a
100 ug
Primary
8515
Mouse