Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrinalpha3/CD49c Mouse anti-Canine, Janelia Fluor 549, Clone: 158A3, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP197691JF549
Description
Integrinalpha3/CD49c Monoclonal antibody specifically detects Integrinalpha3/CD49c in Canine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Integrinalpha3/CD49c | |
158A3 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
Purified | |
ITGA3 | |
158A3 is a mouse monoclonal IgG2a antibody derived by fusion of SP2/0 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a synthetic peptide corresponding to the cytoplasmic domain of the integrin subunit alpha3A including an additio | |
Protein A or G purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Reacts exclusively with the cytoplasmic domain of non-phosphorylated integrin subunit alpha3A. The monoclonal antibody reacts with Human tissues. A broader species reactivity is expected because of the conserved nature of the epitope. |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
antigen identified by monoclonal J143, CD49 antigen-like family member C, CD49c, CD49c antigen, FLJ34631, FLJ34704, FRP-2, Galactoprotein B3, GAP-B3, GAPB3CD49C, integrin alpha-3, integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor), MSK18, VCA-2, very late activation protein 3 receptor, alpha-3 subunit, VL3A, VLA-3 subunit alpha, VLA3a | |
Mouse | |
IgG2a | |
0.1mL | |
Primary | |
3675 | |
Canine, Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title