Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 2/CD49b Antibody (16B4), Biotin, Novus Biologicals™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250144B
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
Integrin alpha 2/CD49b | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
ITGA2 | |
Purified human beta 1 preparation from HT1080 fibrosarcoma cell extract. | |
0.1 mL | |
Cellular Markers, Innate Immunity, Signal Transduction | |
3673 | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
16B4 | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
It is specific to the alpha 2 subunit by relative expression of antigen on various cell lines by Flow Cytomery, and its recognition of affinity purified alpha 2 beta 1 in Dot Blot. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction