Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 2b/CD41 Mouse anti-Human, PerCP, Clone: 2E10C9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237384PCP

Catalog No. NBP237384W

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Integrin alpha 2b/CD41
2E10C9
Flow Cytometry
Purified
ITGA2B
Purified recombinant fragment of human ITGA2B (AA: 45-149) expressed in E. Coli.
Protein A or G purified
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers
Primary
3674
Flow Cytometry
PerCP
PBS with 0.05% Sodium Azide
CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit