Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 4/CD49d Human anti-Human, DyLight™ 550, Clone: Natalizumab, Novus Biologicals™

Human Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275916R

Catalog No. NBP275916R


Add to cart

Description

Description

Integrin alpha 4/CD49d Monoclonal antibody specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Integrin alpha 4/CD49d
Natalizumab
Flow Cytometry
Conjugated, Purified
ITGA4
The RAMOS cell line, a non-EBV releasing cell line, injected into mice.
Protein A purified
Cancer, Cellular Markers, Cytoskeleton Markers, Phospho Specific
Primary
3676.0
Human
Flow Cytometry
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Human
IgG4 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody binds to the alpha 4 subunit of (alpha)4(beta)1 (also known as very late antigen 4 (VLA-4) or CD49d-CD29) and (alpha)4(beta)7 integrin
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit