Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 4/CD49d, Rabbit anti-Human, APC-Cy7, Clone: Natalizumab, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275917APCCY7100UL
Description
Integrin alpha 4/CD49d Monoclonal antibody specifically detects Integrin alpha 4/CD49d in Human Samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 4/CD49d | |
Natalizumab | |
Flow Cytometry | |
Purified | |
ITGA4 | |
The RAMOS cell line, a non-EBV releasing cell line, injected into mice. | |
Protein A purified | |
RUO | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
This antibody binds to the alpha 4 subunit of (alpha)4(beta)1 (also known as very late antigen 4 (VLA-4) or CD49d-CD29) and (alpha)4(beta)7 integrin |
Flow Cytometry | |
APC-Cyanine7 | |
PBS | |
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
Rabbit | |
IgG Kappa | |
100 uL | |
Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
Primary | |
3676 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title