Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 5 beta 1 Human anti-Human, DyLight™ 680, Clone: M200 (Volociximab), Novus Biologicals™

Human Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275909FR

Catalog No. NBP275909FR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Integrin alpha 5 beta 1 Monoclonal antibody specifically detects Integrin alpha 5 beta 1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence.
Specifications

Specifications

Integrin alpha 5 beta 1
M200 (Volociximab)
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Conjugated, Purified
Human
IgG4 Kappa
0.1 mL
Primary
The antibody binds to human (alpha)5(beta)1 integrin with a Kd of 0.367 nM (determined by Biacore), an EC50 of 0.2 nM and an IC50 of 2.3 nM.
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry
DyLight 680
PBS with 0.05% Sodium Azide
ITGA5
Heterodimeric (alpha)5(beta)1Fc fusion protein
Protein A purified
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit