Learn More
Integrin alpha 5 beta 1 Rabbit anti-Human, Alexa Fluor™ 750, Clone: M200 (Volociximab), Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252680AF750
Catalog No. NP22680A70
Add to cart
Description
Description
Integrin alpha 5 beta 1 Monoclonal antibody specifically detects Integrin alpha 5 beta 1 in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunofluorescence.
Specifications
Specifications
Integrin alpha 5 beta 1
M200 (Volociximab)
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
ITGA5
Heterodimeric alpha 5 Beta 1 Fc fusion protein
0.1 mL
Primary
The antibody binds to human (alpha)5(beta)1 integrin with a Kd of 0.367 nM (determined by Biacore), an EC50 of 0.2 nM and an IC50 of 2.3 nM.
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
Human