Learn More
Integrin alpha 6 beta 1 Rabbit anti-Human, Alexa Fluor 350, Clone: 2548B, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB7809U100UG
Catalog No. FAB7809U100UG
Add to cart
This item is not returnable. View return policy
Description
Description
Integrin alpha 6 beta 1 Monoclonal antibody specifically detects Integrin alpha 6 beta 1 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
Integrin alpha 6 beta 1
2548B
Flow Cytometry 0.25-1 ug/10^6 cells
NP_000201
Rabbit
IgG
100 μg
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Human
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Chinese Hamster Ovary cell line, CHO-derived Human Integrin alpha 6 beta 1 heterodimer, Phe24-Ser1012(Integrin alpha 6) and Gln21-Asp728 (Integrin beta1), Accession # NP_000201
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3655