Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 6/CD49f Antibody (450-30A), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064908DL594
Description
Integrin alpha 6/CD49f Monoclonal antibody specifically detects Integrin alpha 6/CD49f in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
Integrin alpha 6/CD49f | |
Monoclonal | |
DyLight 594 | |
CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6 | |
alpha 6 beta 4 integrin purified from A431 cells | |
0.1 mL | |
Signal Transduction | |
3655 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
450-30A | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction