Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 8 Mouse anti-Human, Alexa Fluor 350, Clone: 481709, R&D Systems™

Mouse Monoclonal Antibody

Manufacturer:  R&D Systems FAB6194U100UG

Catalog No. FAB6194U100


Add to cart
This item is not returnable. View return policy

Description

Description

Integrin alpha 8 Monoclonal specifically detects Integrin alpha 8 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Integrin alpha 8
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P53708
ITGA8
E. coli-derived recombinant human Integrin alpha 8 Glu610-Leu800 Accession # P53708
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human Integrin alpha 8 in direct ELISAs and Western blots.
Flow Cytometry
481709
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
integrin alpha-8, integrin, alpha 8, ITGA8
Mouse
IgG2b
RUO
Primary
8516
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)