Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha E/CD103 Rabbit anti-Human, Clone: ITGAE/3904R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$305.50

Specifications

Antigen Integrin alpha E/CD103
Clone ITGAE/3904R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122534
SDP
View Documents
Novus Biologicals
NBP30753020UG
20 μg
Each of 1 for $305.50
Only null left
Add to Cart
 
Description

Description

Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Integrin alpha E/CD103
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Innate Immunity
10 mM PBS with 0.05% BSA
3682
IgG
Protein A or G purified
ITGAE/3904R
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen
Recombinant human Integrin alpha E/CD103 protein fragment (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.