Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha V beta 6 Rabbit anti-Human,Rat, Unconjugated, Clone: EM05201, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Integrin alpha V beta 6
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB110425MI
View Documents
MilliporeSigma
ZRB110425UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB11044XMI
View Documents
MilliporeSigma
ZRB11044X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Integrin alpha V beta 6 Monoclonal antibody specifically detects Integrin alpha V beta 6 in Human,Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

Integrin alpha V beta 6
Unconjugated
Rabbit
Integrin aVb6
Recombinant transmembrane-truncated (DTM) extracellular domains of human integrins avb6.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Rat
ITGA5, ITGB6
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit