Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin beta 2/CD18 Rat anti-Human, Canine, Guinea Pig, PE/Cy5.5, Clone: YFC118.3, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NB200610PECY55
Description
Integrin beta 2/CD18 Monoclonal antibody specifically detects Integrin beta 2/CD18 in Human, Canine, Guinea Pig samples. It is validated for Flow Cytometry.Specifications
Integrin beta 2/CD18 | |
YFC118.3 | |
Flow Cytometry | |
Conjugated, Purified | |
ITGB2 | |
Human neutrophils | |
Protein G purified | |
Apoptosis, Cytokine Research, Immunology, Phospho Specific | |
Primary | |
3689.0 | |
Canine, Guinea Pig, Human |
Flow Cytometry | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
Rat | |
IgG2b | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
This recognises the human CD18 cell surface antigen, the integrin beta2 subunit. CD18 is expressed by the majority of leucocytes. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title