Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 3/CD61 Mouse anti-Human, PE/Cy5.5, Clone: ITGB3/1713, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254562PECY55

Catalog No. N254562PEC5


Add to cart

Description

Description

Integrin beta 3/CD61 Monoclonal antibody specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Integrin beta 3/CD61
ITGB3/1713
Flow Cytometry
Conjugated, Purified
ITGB3
Recombinant human ITGB3 protein
Protein A or G purified
Cancer, Cytoskeleton Markers, Phospho Specific, Signal Transduction
Primary
3690.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit